Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD49e Monoclonal Antibody (5H10-27 (MFR5))
GREENER_CHOICE

Catalog No. PIMA546195
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546195 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546195 Supplier Invitrogen™ Supplier No. MA546195
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

This antibody recognizes an extracellular epitope of murine CD49e, a 135 kDa protein serving as VLA-5 alpha chain, expressed on thymocytes, activated T cells, splenic B cells, and mast cells.

The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49e
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone 5H10-27 (MFR5)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Itga5
Gene Accession No. P11688
Gene Alias 2-phospho-D-glycerate hydro-lyase; alpha enolase like 1; Alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; CD49 antigen-like family member E; Cd49e; C-myc promoter-binding protein; ENO1; ENO1L1; Enolase 1; enolase 1, (alpha); enolase-alpha; epididymis secretory protein Li 17; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; FNRA; HEL-S-17; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin subunit alpha 5; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); ITGA5; Itga5_retired; MBP-1; MBPB1; MPB1; MPB-1; MYC promoter-binding protein 1; NNE; Non-neural enolase; phosphopyruvate hydratase; Plasminogen-binding protein; PPH; tau-crystallin; very late activation protein 5, alpha subunit; VLA5; VLA-5; VLA5A
Gene Symbols Itga5
Host Species Rat
Immunogen C57BL/6 x A/J)F1 murine mast cell line.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16402
Target Species Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.