Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD49f Monoclonal Antibody (NKI-GoH3)
GREENER_CHOICE

Catalog No. PIMA516884
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516884 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516884 Supplier Invitrogen™ Supplier No. MA516884
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. A suggested positive control for immunohistochemical applications is human tonsil.

CDf (Integrin alpha-6, ITGA6) is the integrin alpha chain alpha 6 protein. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. For example, alpha 6 may combine with beta 4 in the integrin referred to as TSP180, or with beta 1 in the integrin VLA-6. Integrins are known to participate in cell adhesion as well as cell-surface mediated signaling. Two transcript variants encoding different isoforms have been found for CDf. Functionally, integrins are cell surface receptors that interact with the extracellular matrix and mediate intracellular signals that affect cellular shape, mobility, and progression through the cell cycle in response to the extracellular matrix. Integrin receptors are composed of alpha and beta subunits, and form structural and functional linkages between the ECM (extra-cellular matrix) and intracellular cytoskeletal linker proteins. Signaling mediated by Intergrin/ECM interactions are also integrated with cellular responses to growth factor signaling to regulate cellular proliferation, cytoskeletal reorganization and other responses necessary for cellular survival. CDf is predominantly expressed by epithelia, and plays a critical structural role in the hemidesmosome. More than 18 alpha and 8 beta subunits with numerous splice variant isoforms have been identified in mammals. Certain integrins can also bind to soluble ligands or to counter-receptors on adjacent cells, such as the intracellular adhesion molecules (ICAMs), resulting in aggregation of cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49f
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone NKI-GoH3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide
Gene Itga6
Gene Accession No. P23229, Q61739
Gene Alias 5033401O05Rik; AI115430; Alpha6p; CD49 antigen-like family member F; CD49f; DKFZp686J01244; FLJ18737; integrin alpha 6; integrin alpha-6; Integrin alpha-6 heavy chain; Integrin alpha-6 light chain; integrin alpha6B; integrin subunit alpha 6; integrin, alpha 6; Itga6; ITGA6B; LOW QUALITY PROTEIN: integrin alpha-6; Processed integrin alpha-6; VLA-6
Gene Symbols Itga6
Host Species Rat
Immunogen BALB/c mouse mammary tumour cells.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100158200, 100856563, 101113656, 101925271, 16403, 3655
Target Species Human, Mouse, Canine, Sheep, Pig, Cynomolgus Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.