Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49f Rat anti-Human, FITC, Clone: GOH3, BD

Catalog No. BDB561893 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561893 25 Tests
BDB555735 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561893 Supplier BD Biosciences Supplier No. 561893
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The GoH3 monoclonal antibody specifically binds to CD49f which is also known as integrin α6 chain. CD49f is a ∽150 kDa type I transmembrane glycoprotein that belongs to the integrin alpha chain family of extracellular matrix and cell adhesion receptors. The integrin α6 subunit associates with the integrin β1 chain (CD29) to form VLA-6 and with the integrin β4 chain (CD104) to form the integrin α6β4 complex, also known as the laminin and kalinin receptor. CD49f is expressed mainly on T cells, monocytes, platelets, epithelial cells, endothelial cells, perineural cells, and trophoblasts of placenta. GoH3 recognizes an extracellular epitope of integrin α6 on human, mouse and bovine cells. GoH3 has been reported to block the binding of integrin α6 to laminin P1 and E8 fragments.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49f
Applications Flow Cytometry
Classification Monoclonal
Clone GoH3
Conjugate FITC
Description ITGA6; IThe GoH3 monoclonal ntegrin alpha-6; Integrin α6 chain; VLA-6; ITA6
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse mammary tumor cells
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Pig
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.