Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD5 Antibody (105), FITC

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290568F
Description
CD5 Monoclonal specifically detects CD5 in Mouse samples. It is validated for ELISA.Specifications
CD5 | |
Monoclonal | |
FITC | |
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD5 extracellular domain (NP_031676.3; Met 1-Pro 371). | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology, Phospho Specific | |
921 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
105 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction