Learn More
CD5 Antibody (4H8E5), Allophycocyanin, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP126707
Catalog No. NBP126707
Only null left
Add to Cart
Description
Description
CD5 Monoclonal specifically detects CD5 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD5
Monoclonal
0.1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry <= 0.2 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
CD5
The immunogen is a peptide made to the amino acid region CD5
0.1 mg
Primary
CD5/Lyt-1, Mr 67 kDa
Store at 4C. Do not freeze.
IgG2a κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
4H8E5
APC
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
Rat
Protein A or G purified
RUO
921
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only