Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD5 Antibody (CD5/54/F6), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234593APCCY7
Description
CD5 Monoclonal specifically detects CD5 in Human, Rat, Porcine, Bovine, Chicken, Equine, Guinea Pig, Monkey, Rabbit samples. It is validated for Flow Cytometry.Specifications
CD5 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
CD5 | |
Recombinant fragment (around aa450-495) of human CD5 protein (intracellular region). (Uniprot: P06127 ) | |
0.1 mL | |
Primary | |
Recognizes a 67kDa transmembrane protein, which is identified as CD5. The CD5 antigen is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. Anti-CD5 is a pan T-cell marker that also reacts with a range of neoplastic B-cells, e.g. chronic lymphocytic leukemia/small lymphocytic lymphoma (CLL/SLL), mantle cell lymphoma, and a subset (∽10%) of diffuse large B-cell lymphoma. CD5 aberrant expression is useful in making a diagnosis of mature T-cell neoplasms. Anti-CD5 detection is diagnostic in CLL/SLL within a panel of other B-cell markers, especially one that includes anti-CD23. Anti-CD5 is also very useful in differentiating among mature small lymphoid cell malignancies. In addition, anti-CD5 can be used in distinguishing thymic carcinoma (+) from thymoma (-). Anti-CD5 does not react with granulocytes or monocytes. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
CD5/54/F6 | |
Flow Cytometry | |
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
Mouse | |
Protein A or G purified | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology, Phospho Specific | |
921.0 | |
Human, Rat, Pig, Bovine, Canine, Chicken, Equine, Guinea Pig, Monkey, Rabbit | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction