Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD5 Antibody (CRIS-1), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234598APC
Description
CD5 Monoclonal specifically detects CD5 in Human, Rhesus Macaque samples. It is validated for Flow Cytometry.Specifications
CD5 | |
Monoclonal | |
APC | |
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
Mouse | |
67 kDa | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Cytokine Research, Immunology | |
921 | |
Human, Primate | |
Purified |
Flow Cytometry | |
CRIS-1 | |
Flow Cytometry | |
CD5 | |
Stimulated human leukocytes | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a 67kDa transmembrane protein, which is identified as CD5 (HLDA I; WS Code T 29HLDA III; WS Code T 530). The CD5 antigen is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. Anti-CD5 is a pan T-cell marker that also reacts with a range of neoplastic B-cells, e.g. chronic lymphocytic leukemia/small lymphocytic lymphoma (CLL/SLL), mantle cell lymphoma, and a subset (∼10%) of diffuse large B-cell lymphoma. CD5 aberrant expression is useful in making a diagnosis of mature T-cell neoplasms. | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction