Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD5 Antibody (MF7-14.5), Novus Biologicals™

Mouse Monoclonal Antibody
$158.00
Specifications
Antigen | CD5 |
---|---|
Clone | MF7-14.5 |
Concentration | 0.025 mg/mL |
Dilution | Flow Cytometry Neat, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017716
![]() |
Novus Biologicals
NB100653770.025ML |
0.025 mL |
Each for $158.00
|
|
|||||
NB10065377
![]() |
Novus Biologicals
NB10065377 |
0.1 mL | N/A | N/A | N/A | ||||
Description
CD5 Monoclonal specifically detects CD5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
CD5 | |
0.025 mg/mL | |
Immunohistochemistry, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cytokine Research, Immunology | |
P06127 | |
921 | |
Made to Human CD5 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MF7-14.5 | |
Flow Cytometry Neat, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
CD5 | |
IgG2a | |
Protein A or G purified | |
NB100-65377 recognises the CD5 cell surface glycoprotein, a 67kD antigen expressed by T lymphocytes and by a subset of B lymphocytes. CD5 is also expressed by tumour cells in cases of B-cell Chronic Lymphocytic Leukaemia (B-CLL) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title