Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD5 Monoclonal Antibody (CG16), Biotin
GREENER_CHOICE

Catalog No. PIMA517785
Change view
Click to view available options
Quantity:
100 μg
300 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517785 100 μg
PIMA517786 300 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA517785 Supplier Invitrogen™ Supplier No. MA517785
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD5 Monoclonal Antibody for Flow

CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter's Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD5
Applications Flow Cytometry
Classification Monoclonal
Clone CG16
Concentration 0.1 mg/mL
Conjugate Biotin
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene Cd5
Gene Accession No. P13379
Gene Alias alanyl (membrane) aminopeptidase; alanyl aminopeptidase; alanyl aminopeptidase, membrane; aminopeptidase M; Aminopeptidase N; ANPEP; AP-M; APN; AP-N; CD antigen CD5; CD13; CD28; CD28 antigen (Tp44); CD28 molecule; Cd5; CD5 antigen; CD5 antigen (p56 62); CD5 antigen (p56-62); CD5 antigen p56-62; CD5 molecule; CD5 protein; CD74 antigen, invariant polypeptide of major; cell surface protein; costimulatory molecule B7 receptor CD28; fCD5; GP150; hAPN; LAP1; LEU1; Ly-1; Ly12; Ly-12; LyA; Ly-A; Lymphocyte antigen 1; Lymphocyte antigen CD5; lymphocyte antigen T1/Leu-1; Lyt-1; membrane alanyl aminopeptidase; microsomal aminopeptidase; Myeloid plasma membrane glycoprotein CD13; P150; Pan T cell; PEPN; T1; T-cell costimulatory molecule CD28; T-cell surface glycoprotein CD5; T-cell-specific surface glycoprotein CD28; unnamed protein product
Gene Symbols Cd5
Host Species Mouse
Immunogen C3H.CE - Ly 1.2: DS.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12507
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.