Learn More
Invitrogen™ CD5 Monoclonal Antibody (CVS5), PE

Description
Mouse anti Horse CD5 antibody, clone CVS5 recognizes the equine CD5 antigen, a single-pass type I membrane protein with an approximate molecular weight of 69 kDa. Reconstitute by adding 1 mL distilled water.
Specifications
Specifications
| Antigen | CD5 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | CVS5 |
| Concentration | Conc. Not Determined |
| Conjugate | PE |
| Formulation | PBS with 5% sucrose, 1% BSA and 0.09% sodium azide |
| Gene | Cd5 |
| Gene Accession No. | 0 |
| Gene Alias | alanyl (membrane) aminopeptidase; alanyl aminopeptidase; alanyl aminopeptidase, membrane; aminopeptidase M; Aminopeptidase N; ANPEP; AP-M; APN; AP-N; CD antigen CD5; CD13; CD28; CD28 antigen (Tp44); CD28 molecule; Cd5; CD5 antigen; CD5 antigen (p56 62); CD5 antigen (p56-62); CD5 antigen p56-62; CD5 molecule; CD5 protein; CD74 antigen, invariant polypeptide of major; cell surface protein; costimulatory molecule B7 receptor CD28; fCD5; GP150; hAPN; LAP1; LEU1; Ly-1; Ly12; Ly-12; LyA; Ly-A; Lymphocyte antigen 1; Lymphocyte antigen CD5; lymphocyte antigen T1/Leu-1; Lyt-1; membrane alanyl aminopeptidase; microsomal aminopeptidase; Myeloid plasma membrane glycoprotein CD13; P150; Pan T cell; PEPN; T1; T-cell costimulatory molecule CD28; T-cell surface glycoprotein CD5; T-cell-specific surface glycoprotein CD28; unnamed protein product |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.