Learn More
CD5 Mouse anti-Human, APC, Clone: UCHT2, BD
Mouse Monoclonal Antibody
$162.70 - $404.50
Specifications
| Antigen | CD5 |
|---|---|
| Clone | UCHT2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
Description
The UCHT2 monoclonal antibody specifically binds to CD5. CD5 is a 67 kDa single-chain, type 1 transmembrane glycoprotein expressed on most thymocytes, the majority of peripheral T lymphocytes and a subpopulation of B cells. CD72 has been shown to be the natural ligand for CD5. CD5+ B cells produce polyreactive antibodies (mostly IgM).
Specifications
| CD5 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| UCHT2 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human T cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.