Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5 Mouse anti-Human, Brilliant Violet 786, Clone: L17F12, BD Optibuild™

Catalog No. BDB742553 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742553 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742553 Supplier BD Biosciences Supplier No. 742553
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The L17F12 monoclonal antibody specifically binds to CD5 which is also known as Lymphocyte antigen T1 (T1) or LEU1/Leu-1. CD5 is an ñ67 kDa single-chain, type 1 transmembrane glycoprotein expressed on most thymocytes, the majority of peripheral T lymphocytes and a subpopulation of B cells. CD5+ B cells produce polyreactive antibodies (mostly IgM). This CD5-specific antibody also reacts with many T-cell leukemias and lymphomas and most Ig+ B-cell chronic lymphoblastic leukemia (B-CLL) cells. CD72 has been shown to be the natural ligand for CD5.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD5
Classification Monoclonal
Clone L17F12
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias LEU1, Leu-1, Lymphocyte antigen T1, T1, LY1, Tp67
Host Species Mouse
Immunogen Human T-cell acute lymphoblastic leukemia (T-ALL) cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.