Learn More
CD5 Mouse anti-Human, FITC, Clone: UCHT2, BD
Mouse Monoclonal Antibody
$104.00 - $230.00
Specifications
| Antigen | CD5 |
|---|---|
| Clone | UCHT2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
BDB561896
|
BD Biosciences
561896 |
25 Tests |
Each for $104.00
|
|
|||||
|
BDB555352
|
BD Biosciences
555352 |
100 Tests |
Each for $230.00
|
|
|||||
Description
The UCHT2 monoclonal antibody specifically binds to CD5. CD5 is a 67 kDa single-chain, type 1 transmembrane glycoprotein expressed on most thymocytes, the majority of peripheral T lymphocytes and a subpopulation of B cells. CD72 has been shown to be the natural ligand for CD5. CD5+ B cells produce polyreactive antibodies (mostly IgM).
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD5 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| UCHT2 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human T cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.