Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD5 Rat anti-Mouse, R-PE, Clone: B19.1, Southern Biotech™

Rat Monoclonal Antibody
$235.50 - $382.00
Specifications
| Antigen | CD5 |
|---|---|
| Clone | B19.1 |
| Concentration | 0.1mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
| CD5 | |
| 0.1mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| IgG2a | |
| Store at 2-8°C; Protect from light; Do not freeze |
| B19.1 | |
| Flow Cytometry | |
| R-PE | |
| RUO | |
| Supplied in PBS/NaN3 and a stabilizing agent | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title