Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD50 (ICAM-3) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119214
This item is not returnable.
View return policy
Description
CD50 (ICAM-3) Polyclonal Antibody for Western Blot, IHC (P)
CD50 (intracellular adhesion molecule 3, ICAM-3) is a transmembrane glycoprotein expressed by leukocytes, that serves as a counter-receptor for the lymphocyte function-associated antigen (LFA)-1 integrin. Besides functioning as an adhesive molecule that mediates e.g. the contact between T cells and antigen presenting cells, ICAM-3 regulates affinity of LFA-1 for ICAM-1 and induces T cell activation and proliferation. ICAM-3 plays an essential role in the initiation of the immune response both on T cells and antigen presenting cells and interacts also with CD209 (dendritic cell-specific ICAM-3-grabbing nonintegrin, DC-SIGN), a C-type lectin of dendritic cells and macrophages; this process is involved in dialogue between dendritic cells and granulocytes.
Specifications
CD50 (ICAM-3) | |
Polyclonal | |
Unconjugated | |
ICAM3 | |
CD50; CDw50; ICAM3; ICAM-3; ICAM-R; Intercellular adhesion molecule 3; intercellular adhesion molecule-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3385 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P32942 | |
ICAM3 | |
Recombinant protein Intercellular Adhesion Molecule 3. The antigen corresponds to amino acid range 46-197 of the target protein. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction