Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD50 Mouse anti-Human, Brilliant Violet 421, Clone: TU41, BD Optibuild™

Catalog No. BDB742303 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742303 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742303 Supplier BD Biosciences Supplier No. 742303
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TU41 monoclonal antibody specifically binds to the intracellular adhesion molecule-3 (ICAM-3) which is also known as CD50. ICAM-3 is a 110-120 kDa type I membrane glycoprotein expressed on all leukocytes (platelets and erythrocytes are ICAM-3-negative). ICAM-3 is also expressed on epidermal Langerhans cells and endothelial cells. It predominantly interacts with LFA-1 (CD11a/CD18) without requiring activation as the initial dendritic cell-T lymphocyte interaction. ICAM-3 is an important LFA-1 ligand that functions in the initiation of the immune/inflammatory response and also plays a role in chemotaxis.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD50
Classification Monoclonal
Clone TU41
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ICAM-3, ICAM3, Intercellular adhesion molecule 3
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.