Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD50 Mouse anti-Human, BUV395, Clone: TU41, BD Optibuild™

Catalog No. BDB742307 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742307 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742307 Supplier BD Biosciences Supplier No. 742307
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TU41 monoclonal antibody specifically binds to the intracellular adhesion molecule-3 (ICAM-3) which is also known as CD50. ICAM-3 is a 110-120 kDa type I membrane glycoprotein expressed on all leukocytes (platelets and erythrocytes are ICAM-3-negative). ICAM-3 is also expressed on epidermal Langerhans cells and endothelial cells. It predominantly interacts with LFA-1 (CD11a/CD18) without requiring activation as the initial dendritic cell-T lymphocyte interaction. ICAM-3 is an important LFA-1 ligand that functions in the initiation of the immune/inflammatory response and also plays a role in chemotaxis.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD50
Classification Monoclonal
Clone TU41
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ICAM-3, ICAM3, Intercellular adhesion molecule 3
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.