Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD51/CD61 Monoclonal Antibody (23C6), PE
GREENER_CHOICE

Catalog No. PIMA544118
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544118 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544118 Supplier Invitrogen™ Supplier No. MA544118
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope on CD51/CD61 complex, that is expressed mainly on human osteoclasts, but also e.g. on placenta, or melanoma cell lines. The epitope is native and sensitive to fixation. In chicken this antibody can be used to selectively identify the thrombocytes.

CD51 and CD61 molecules (alpha V and beta 3 integrins) form a complex. This complex binds vitronection at the RGD sequence and can also bind fibrinogen, von Willebrand factor, thrombospondin, fibronectin, osteopontin and collagen. Reacts with osteoclasts, placenta, melanoma cell lines and weakly with platelets.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD51/CD61
Applications Flow Cytometry
Classification Monoclonal
Clone 23C6
Concentration 0.08 mg/mL
Conjugate PE
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene ITGB3
Gene Accession No. P05106, P06756, P26008
Gene Alias 1110004F14Rik; 2610028E01Rik; alpha v integrin; antigen CD61; antigen identified by monoclonal antibody L230; BDPLT16; BDPLT2; Cd51; CD61; D430040G12Rik; Dnmt3l-ps1 pseudogene; GP3A; GPIIIa; GT; INGRB3; integrin; integrin alpha V; integrin alpha-5 beta-3; integrin alpha-V; Integrin alpha-V heavy chain; Integrin alpha-V light chain; integrin alphaVbeta3; Integrin beta; integrin beta 3; integrin beta 3 (Cd61); Integrin beta3; integrin beta-3; integrin subunit alpha V; integrin subunit beta 3; integrin, alpha V; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGAV; Itgb3; MSK8; platelet gpIIIa; platelet membrane glycoprotein IIIa; Vitronectin receptor; vitronectin receptor alpha polypeptide (VNRA); vitronectin receptor alpha subunit; vitronectin receptor subunit alpha; VNRA; VTNR
Gene Symbols ITGAV, ITGB3
Host Species Mouse
Immunogen A cell suspension containing osteoclasts from osteoclastomas.
Purification Method Size-exclusion chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3685, 3690, 374209, 396420
Target Species Human, Chicken
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.