Learn More
Invitrogen™ CD51 (Integrin alpha V) Monoclonal Antibody (RMV-7)

Description
This antibody recognizes murine CD51, approximately a 140 kDa alpha subunit of the vitronectin receptor, which is otherwise known as the integrin alpha v chain. CD51 can form heterodimers at the cell surface with a variety of beta integrins including CD29 and CD61.
Specifications
Specifications
| Antigen | CD51 (Integrin alpha V) |
| Applications | Flow Cytometry, Functional Assay, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | RMV-7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | ITGAV |
| Gene Accession No. | P43406 |
| Gene Alias | 1110004F14Rik; 2610028E01Rik; alpha v integrin; antigen identified by monoclonal antibody L230; Cd51; CD61; D430040G12Rik; Dnmt3l-ps1 pseudogene; GP3A; GPIIIa; integrin alpha V; integrin alpha-V; Integrin alpha-V heavy chain; Integrin alpha-V light chain; integrin alphaVbeta3; integrin subunit alpha V; integrin, alpha V; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); ITGAV; ITGB3; MSK8; Vitronectin receptor; vitronectin receptor alpha polypeptide (VNRA); vitronectin receptor subunit alpha; VNR; VNRA; VTNR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.