Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD52, anti-Human, Clone: Campath-1H, Abnova™
Monoclonal Antibody
Supplier: Abnova Corporation RAB00044
Specifications
CD52 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.2 (0.02% Proclin 300). | |
CDW52 | |
Original antibody is raised against human lymphocytes. | |
RUO | |
1043 | |
Store at 4°C for up to 3 months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Flow Cytometry | |
Campath-1H | |
Flow Cytometry (1 ug/mL) The optimal working dilution should be determined by the end user. | |
CD52 molecule | |
CD52 | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction