Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD52 Antibody (4C8), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311701AF405
Description
CD52 Monoclonal antibody specifically detects CD52 in Human samples. It is validated for Flow CytometrySpecifications
CD52 | |
Monoclonal | |
Alexa Fluor 405 | |
Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5 | |
Human lymphocytes | |
0.1 mL | |
Cancer, Immunology | |
1043 | |
Store at 4°C in the dark. | |
IgG3 |
Flow Cytometry | |
4C8 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction