Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD52 Antibody (Campath-1H), DyLight 550, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275896R
Description
CD52 Monoclonal specifically detects CD52 in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
CD52 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD52 | |
Human lymphocytes | |
0.1 mL | |
Primary | |
Human CD52 antigen, also known as CAMPATH-1. CD52 is expressed at high density by lymphocytes, monocytes, eosinophils, thymocytes and macrophages. It is expressed by most lymphoid derived malignancies, although expression on myeloma cells is variable. | |
Store at 4°C in the dark. |
ELISA, Immunohistochemistry, Immunofluorescence | |
Campath-1H | |
ELISA, Immunohistochemistry, Immunofluorescence | |
Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5 | |
Human | |
Protein A purified | |
Cancer, Immunology | |
1043.0 | |
Human, Cynomolgus Monkey, Rhesus Macaque | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction