Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD52 Antibody (Campath-1H), PerCP, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275896PCP
Description
CD52 Monoclonal antibody specifically detects CD52 in Human, Cynomolgus Monkey, Macaque samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ImmunofluorescenceSpecifications
| CD52 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Cynomolgus Monkey, Macaque | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Campath-1H | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay, Immunofluorescence | |
| Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5 | |
| Human lymphocytes | |
| 0.1 mL | |
| Cancer, Immunology | |
| 1043 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction