Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD52 Antibody (SR2268), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321870100UL
Description
CD52 Monoclonal antibody specifically detects CD52 in Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
CD52 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
SR2268 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5 | |
A synthesized peptide derived from human CD52 (Uniprot #: Q64389) | |
100 μg | |
Cancer, Immunology | |
1043 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction