Learn More
Invitrogen™ CD52 Monoclonal Antibody (11/305/44)

Description
Mouse anti Pig CD52, clone 11/305/44 is a monoclonal antibody recognizing the porcine homologue of human CD52, a approximately 19 kDa antigen expressed by mature lymphocytes, monocytes and dendritic cells.
Specifications
Specifications
| Antigen | CD52 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 11/305/44 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | CD52 |
| Gene Accession No. | 0 |
| Gene Alias | AI463198; B7; B7-Ag; cambridge pathology 1 antigen; CAMPATH-1; CAMPATH-1 antigen; Cd52; CD52 antigen; CD52 antigen (CAMPATH-1 antigen); CD52 molecule; CDW52; CDW52 antigen (CAMPATH-1 antigen); CLS1; Epididymal secretory protein E5; epididymis secretory sperm binding protein Li 171mP; he5; HEL-S-171mP; human epididymis-specific protein 5; Lymphocyte differentiation antigen B7; Mb7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.