Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD52 Monoclonal Antibody (CF1D12), Super Bright™ 702, eBioscience™
GREENER_CHOICE

Catalog No. 67052942
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-052-942 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-052-942 Supplier Invitrogen™ Supplier No. 67052942
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This CF1D12 monoclonal antibody reacts with human CD52, a cell surface glycoprotein expressed in the immune and reproductive systems. CF1D12 is thought to recognize the carbohydrate region of CD52, not the protein core of the molecule. Tissue-specific modifications of the glycan structure may alter epitope recognition in certain applications. For example, this CF1D12 clone has been reported to recognize human sperm glycoforms of CD52 by western blot, but not by flow cytometry. The immunogen used to generate this antibody was human CD52. Applications Reported: This CF1D12 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CF1D12 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interac...

CD52, also known as CAMPATH-1 antigen, is a glycoprotein that is expressed on the surface of various immune cells, including T and B lymphocytes, natural killer cells, and monocytes. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including multiple sclerosis, chronic lymphocytic leukemia, and autoimmune disorders. CD52 is the target of alemtuzumab, a monoclonal antibody that is used to treat these conditions by depleting immune cells that express CD52.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD52
Applications Flow Cytometry
Classification Monoclonal
Clone CF1D12
Concentration 5 μL/Test
Conjugate Super Bright 702
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CD52
Gene Accession No. P31358
Gene Alias AI463198; B7; B7-Ag; cambridge pathology 1 antigen; CAMPATH-1; CAMPATH-1 antigen; Cd52; CD52 antigen; CD52 antigen (CAMPATH-1 antigen); CD52 molecule; CDW52; CDW52 antigen (CAMPATH-1 antigen); CLS1; Epididymal secretory protein E5; epididymis secretory sperm binding protein Li 171mP; he5; HEL-S-171mP; human epididymis-specific protein 5; Lymphocyte differentiation antigen B7; Mb7
Gene Symbols CD52
Host Species Mouse
Immunogen Human CD52.
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1043
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.