Learn More
Invitrogen™ CD52 Monoclonal Antibody (YTH34.5), FITC
Rat Monoclonal Antibody
$313.00 - $723.00
Specifications
Antigen | CD52 |
---|---|
Clone | YTH34.5 |
Concentration | 0.1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA517000
|
Invitrogen™
MA517000 |
100 μg |
Each for $723.00
|
|
|||||
PIMA517001
|
Invitrogen™
MA517001 |
25 μg |
Each for $313.00
|
|
|||||
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Human CD52 antibody, clone YTH34.5 reacts with the human CD52 antigen, also known as CAMPATH-1. Rat anti Human CD52 antibody, clone YTH34.5 reacts with the human CD52 antigen, also known as CAMPATH-1.
CD52, also known as CAMPATH-1 antigen, is a glycoprotein that is expressed on the surface of various immune cells, including T and B lymphocytes, natural killer cells, and monocytes. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including multiple sclerosis, chronic lymphocytic leukemia, and autoimmune disorders. CD52 is the target of alemtuzumab, a monoclonal antibody that is used to treat these conditions by depleting immune cells that express CD52.Specifications
CD52 | |
0.1 mg/mL | |
Flow Cytometry | |
FITC | |
Rat | |
RUO | |
PBS with 1% BSA and 0.09% sodium azide | |
P31358 | |
1043, 714429 | |
human lymphocytes. | |
Antibody |
YTH34.5 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Monoclonal | |
Liquid | |
IgG2b | |
Human, Rhesus Macaque | |
CD52 | |
AI463198; B7; B7-Ag; cambridge pathology 1 antigen; CAMPATH-1; CAMPATH-1 antigen; Cd52; CD52 antigen; CD52 antigen (CAMPATH-1 antigen); CD52 molecule; CDW52; CDW52 antigen (CAMPATH-1 antigen); CLS1; Epididymal secretory protein E5; epididymis secretory sperm binding protein Li 171mP; he5; HEL-S-171mP; human epididymis-specific protein 5; Lymphocyte differentiation antigen B7; Mb7 | |
CD52 | |
Primary | |
Protein A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.