Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD52 Mouse anti-Human, BV 421, Clone: 4C8, BD Horizon™
Mouse Monoclonal Antibody
$268.00 - $468.00
Specifications
Antigen | CD52 |
---|---|
Clone | 4C8 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 421 |
Description
CD52 Monoclonal antibody specifically detects CD52 in Human samples. It is validated for Flow CytometrySpecifications
CD52 | |
Flow Cytometry | |
Brilliant Violet 421 | |
Mouse | |
Human | |
Cambridge pathology 1 Ag; CAMPATH-1; Epididymal secretory protein E5; HE5 | |
IgG3 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
4C8 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human T Lymphocytes | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title