Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD53 Antibody (161-2 (53/2)), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312077G
Description
CD53 Monoclonal antibody specifically detects CD53 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
CD53 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG κ |
Flow Cytometry, Immunofluorescence | |
161-2 (53/2) | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53 | |
This antibody was raised by immunizng mice with stimulated human leukocytes. | |
0.1 mL | |
Immunology | |
963 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction