Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD53 Antibody (425514), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4624PECY7
Description
CD53 Monoclonal antibody specifically detects CD53 in Human samples. It is validated for Flow CytometrySpecifications
CD53 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
425514 | |
Flow Cytometry | |
antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53 | |
NS0 mouse myeloma cell line transfected with human CD53, Met1-Leu219, Accession # P19397 | |
0.1 mL | |
Immunology | |
963 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction