Learn More
Invitrogen™ CD53 Chimeric Recombinant Rabbit Monoclonal Antibody (161-2 (53/2))
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA548272
Description
Specificity: This antibody recognizes human CD53. It does not react with the baboon and horse antigen. Cd53 is a member of tetraspanin family and is involved in cellular activation as part of a signal transduction complex involving other membrane glycoproteins.
CD53 is a tetraspanin family transmembrane glycoprotein expressed in the lymphoid-myeloid lineage. This molecule has been reported to form complexes with other leukocyte surface proteins such as CD2, CD19, CD21, MHC II, VLA-4 or tetraspanins CD37, CD81 and CD82, thus probably modulating various signaling processes. CD53 is involved in radioresistancy of tumor cells and its triggering has anti-apoptotic effect. In thymus, CD53 is up-regulated in response to positive selection signals during T cell development, and is strongly expressed upon macrophage exposure to bacterial lipopolysaccharide, whereas stimulation of neutrophils results in down-regulation of CD53 expression.
Specifications
CD53 Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P19397 | |
Cd53 | |
This antibody was raised by immunizng mice with stimulated human leukocytes. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG κ |
Flow Cytometry, Functional Assay, Immunocytochemistry | |
161-2 (53/2) | |
Unconjugated | |
Cd53 | |
AI323659; antigen MOX44 identified by monoclonal antibody MRC-OX44; Cd53; CD53 antigen; CD53 glycoprotein; CD53 molecule; CD53 tetraspan antigen; cell surface antigen; cell surface glycoprotein CD53; Leukocyte antigen MRC OX-44; leukocyte surface antigen CD53; MOX44; OX44; Ox-44; tetraspanin-25; transmembrane glycoprotein; TSPAN25; tspan-25 | |
Rabbit | |
Protein A | |
RUO | |
963 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.