Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD53 Monoclonal Antibody (1H2C7)
GREENER_CHOICE

Catalog No. FERMA548414
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548414 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548414 Supplier Invitrogen™ Supplier No. MA548414
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD53 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

CD53 is a tetraspanin family transmembrane glycoprotein expressed in the lymphoid-myeloid lineage. This molecule has been reported to form complexes with other leukocyte surface proteins such as CD2, CD19, CD21, MHC II, VLA-4 or tetraspanins CD37, CD81 and CD82, thus probably modulating various signaling processes. CD53 is involved in radioresistancy of tumor cells and its triggering has anti-apoptotic effect. In thymus, CD53 is up-regulated in response to positive selection signals during T cell development, and is strongly expressed upon macrophage exposure to bacterial lipopolysaccharide, whereas stimulation of neutrophils results in down-regulation of CD53 expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD53
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone 1H2C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Cd53
Gene Accession No. P19397
Gene Alias AI323659; antigen MOX44 identified by monoclonal antibody MRC-OX44; Cd53; CD53 antigen; CD53 glycoprotein; CD53 molecule; CD53 tetraspan antigen; cell surface antigen; cell surface glycoprotein CD53; Leukocyte antigen MRC OX-44; leukocyte surface antigen CD53; MOX44; OX44; Ox-44; tetraspanin-25; transmembrane glycoprotein; TSPAN25; tspan-25
Gene Symbols Cd53
Host Species Mouse
Immunogen Purified recombinant fragment of human CD53 (AA: extra mix) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 963
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.