Learn More
CD53 Mouse anti-Human, Brilliant Violet 650, Clone: HI29, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 742965
Description
The HI29 monoclonal antibody specifically recognizes CD53. CD53 is a 35-42 kDa type III transmembrane glycoprotein that is expressed on all leucocytes, including plasma cells, but not on platelets, erythrocytes, and non-hematopoietic tissues. CD53 mediates signal transduction in monocytes and B cells. It has the broadest reactivity of all pan-leukocyte reagents used in immunohistology applications.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
Specifications
| CD53 | |
| HI29 | |
| Brilliant Violet 650 | |
| tetraspanin-25, TSPAN25, tspan-25, MOX44 | |
| Human Peripheral Blood Mononuclear Cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.