Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD53 Rabbit anti-Human, Cynomolgus Monkey, Clone: 104, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | CD53 |
---|---|
Clone | 104 |
Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate |
Applications | Western Blot, ELISA, Immunoprecipitation |
Classification | Monoclonal |
Description
CD53 Monoclonal specifically detects CD53 in Human, Cynomolgus Monkey samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
CD53 | |
Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate | |
Monoclonal | |
Purified | |
RUO | |
Human, Cynomolgus Monkey | |
antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53 | |
Recombinant Cynomolgus CD53 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
104 | |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Immunology | |
0.2 um filtered solution in PBS | |
963 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title