Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD54/ICAM-1 Ab-4, Mouse Monoclonal Antibody

Catalog No. MS1094S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1094S1 500 μL Concentrated
MS1094S 1 mL Concentrated
MS1094R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1094S1 Supplier Epredia™ Supplier No. MS1094S1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD54/ICAM-1 Ab-4, Mouse Monoclonal Antibody.

ICAM-1 (intercellular adhesion molecule) has 7 potential N-linked glycosylation sites. It is a single chain glycoprotein of Ig supergene family, present on unstimulated endothelial cells (EC) and on a variety of other cell types including activated fibroblasts, EC, macrophages, and lymphocytes. ICAM-1 mediates cell adhesion by binding to integrins CD11a/CD18 (leukocyte adhesion molecule, LFA-1) and to CD11b/CD18 (Mac-1).

Host Species: Mouse

Clone: 23G12

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Recombinant protein encoding part of the external domain of human CD54

Molecular Weight: 85-115 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting (Not suitable)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD54 Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 23G12
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding part of the external domain of human CD54
Purification Method Tonsil
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.