Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD54 (ICAM-1) Monoclonal Antibody (eBioKAT-1 (KAT-1, KAT1)), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5012651
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-126-51 100 μg
50-126-52 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-126-51 Supplier Invitrogen™ Supplier No. 14054282
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The eBioKAT-1 monoclonal antibody reacts with mouse CD54 (ICAM-1), which is a 95 kDa member of the immunoglobulin superfamily. CD54 is expressed at low levels on leukocytes and high endothelial venules, and expression increases in response to inflammatory cytokines. ICAM-1 binds to LFA-1 and this interaction is required for the transendothelial migration of T cells. ICAM-1-deficient mice are viable, however the migration of leukocytes to sites of inflammation is reduced leading to impaired immune and inflammatory responses. Based on the regulated expression of ICAM-1, it has been suggested that ICAM-1 may increase leukocyte extravasation at sites of inflammation, whereas the constitutively high expression of ICAM-2 mediates leukocyte traffic into non-inflamed tissue. The eBioKAT-1 monoclonal antibody recognizes a different epitope than the YN1/1.7.4 monoclonal antibody. Applications Reported: This eBioKAT-1 (KAT-1, KAT1) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunohistology staining of frozen tissue sections, and immunohistology staining of paraffin embedded tissue sections. (Please use Functional Grade purified eBioKAT-1 (KAT-1, KAT1), cat. 16-0542, in functional assays.). Applications Tested: This eBioKAT-1 (KAT-1, KAT1) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test.

ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD54 (ICAM-1)
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone eBioKAT-1 (KAT-1, KAT1), KAT1)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene ICAM1
Gene Accession No. P13597
Gene Alias BB2; CD54; cell surface glycoprotein P3.58; Human rhinovirus receptor; ICAM; Icam1; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; intercellular adhesion molecule-1; intercellular adhesion molecule-1 precursor; Ly 47; Ly-47; major group rhinovirus receptor; MALA2; MALA-2; MyD10; P3.58; sCD54; soluble CD54; Surface antigen of activated B cells
Gene Symbols ICAM1
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15894
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.