Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD54 (ICAM-1) Monoclonal Antibody (HA58), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD54 (ICAM-1) |
---|---|
Clone | HA58 |
Concentration | 0.4 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H035T03Y03A
![]() |
Invitrogen™
H035T03Y03A |
100 Tests |
Each for $430.00
|
|
|||||
H035T02Y03A
![]() |
Invitrogen™
H035T02Y03A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD54 (ICAM-1) Monoclonal antibody specifically detects CD54 (ICAM-1) in Human samples. It is validated for Flow CytometrySpecifications
CD54 (ICAM-1) | |
0.4 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P05362 | |
3383 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
HA58 | |
Flow Cytometry | |
NovaFluor Yellow 610 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
BB2; CD54; cell surface glycoprotein P3.58; Human rhinovirus receptor; ICAM; Icam1; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; intercellular adhesion molecule-1; intercellular adhesion molecule-1 precursor; Ly 47; Ly-47; major group rhinovirus receptor; MALA2; MALA-2; MyD10; P3.58; sCD54; soluble CD54; Surface antigen of activated B cells | |
IgG1 κ | |
Affinity chromatography | |
ICAM1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title