Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD54 (ICAM-1) Rat anti-Mouse, PE, Clone: YN1/1.7.4 (also known as YN1/1.7), BD Pharmingen™
Rat Monoclonal Antibody
$84.00 - $246.00
Specifications
Antigen | CD54 (ICAM-1) |
---|---|
Clone | YN1/1.7.4 (also known as YN1/1.7) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD54 (ICAM-1) Monoclonal antibody specifically detects CD54 (ICAM-1) in Mouse samples. It is validated for Flow cytometry (Routinely Tested)Specifications
CD54 (ICAM-1) | |
0.2 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse NS-1 Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
YN1/1.7.4 (also known as YN1/1.7) | |
Flow Cytometry | |
PE | |
Rat | |
Mouse | |
Icam-1; Icam1; intercellular adhesion molecule 1; Ly-47; MALA-2; myD10 | |
IgG2b κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title