Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD54 Mouse, anti-Rat, R718, Clone: 1A29, BD OptiBuild™

Catalog No. BDB751649 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751649 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751649 Supplier BD Biosciences Supplier No. 751649
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1A29 monoclonal antibody specifically recognizes ICAM-1 which is also known as CD54. ICAM-1 is an 85 kDa type I transmembrane glycoprotein that is encoded by Icam1 (Intercellular adhesion molecule 1). It is expressed on vascular endothelium in lymphoid tissues, thymic stromal cells, peripheral blood monocytes, peritoneal macrophages and mast cells, dendritic cells, and weakly on peripheral lymphocytes and thymocytes. ICAM-1 is a ligand for LFA-1 (CD11a/CD18) and Mac-1 (CD11b/CD18). Its expression is upregulated on activated lymphocytes and endothelial cells. 1A29 mAb inhibits Phorbol 12-Myristate 13-Acetate (PMA)-induced aggregation of phytohemagglutinin (PHA)-stimulated splenic blasts, as well as the adhesion of mitogen-stimulated blasts to high endothelial venule (HEV) cells and purified ICAM-1. The 1A29 antibody can reportedly inhibit leucocyte infiltration in in vivo systems, blocks induction of experimental allergic encephalomyelitis, and reduces NK-cell adhesion to tumor cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD54
Applications Flow Cytometry
Classification Monoclonal
Clone 1A29
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.