Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD55/DAF Antibody (028), DyLight 755

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289269IR
Description
CD55/DAF Monoclonal specifically detects CD55/DAF in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CD55/DAF | |
Monoclonal | |
DyLight 755 | |
CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD55/DAF (NP_000565.1; Met 1-Ser 353). | |
0.1 mL | |
Immunology | |
1604 | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
28 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human CD97 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction