Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD55/DAF Antibody (143-30), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247964MFV500
Description
CD55/DAF Monoclonal antibody specifically detects CD55/DAF in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface)Specifications
CD55/DAF | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
143-30 | |
Flow Cytometry, Flow (Cell Surface) | |
CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC | |
PHA stimulated human CD55/DAF (Uniprot: P08174 ) | |
0.1 mL | |
Immunology | |
1604 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction