Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD55/DAF Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241295
Description
CD55/DAF Polyclonal specifically detects CD55/DAF in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
CD55/DAF | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
CD55 | |
Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human DAF. The immunogen is located within amino acids 360 - 410 of DAF. Amino Acid Squence: DRSSLQSRTPGFKRSFH | |
Affinity Purified | |
RUO | |
Primary | |
DAF antibody is human specific. At least two isoforms of DAF are known to exist; this antibody will only detect the longest isoform. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml, Immunofluorescence 20 ug/ml | |
CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC | |
Rabbit | |
49 kDa | |
0.1 mg | |
Immunology | |
1604 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction