Learn More
Invitrogen™ CD55 Monoclonal Antibody (583905)

Description
In direct ELISAs, no cross-reactivity with recombinant human CD55 or recombinant mouse CD97 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | CD55 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 583905 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CD55 |
| Gene Accession No. | Q61475 |
| Gene Alias | CD55; CD55 antigen; CD55 molecule (Cromer blood group); CD55 molecule, decay accelerating factor for complement; CD55 molecule, decay accelerating factor for complement (Cromer blood group); Cd55a; Complement decay-accelerating factor; complement decay-accelerating factor, GPI-anchored; complement-glycosylphosphatidylinositol; CR; CROM; Cromer blood group; Daf; Daf1; DAF-GPI; decay accelarating factor 1; decay accelerating factor 1; decay accelerating factor GPI-form; decay-accelarating factor; glycosylphophatidylinositol-anchored form; GPI anchor addition signal; GPI-DAF; RP11-357P18.1; TC |
| Show More |
Frequently Asked Questions (FAQs)
Unfortunately, this antibody has not been internally verified for ICC/IF, and therefore we cannot guarantee that it will work for this application.
Alternatively, we offer CD55 Recombinant Rabbit Monoclonal Antibody (76) (Cat. No. MA5-29678) and CD55 Polyclonal Antibody (Cat. No. PA5-88462) that target mouse and have been verified for ICC/IF.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.