Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD55 Mouse anti-Human, PE, Clone: IA10, BD

Catalog No. BDB341030 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
50 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB341030 50 Tests
BDB561901 25 Tests
BDB555694 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BDB341030 Supplier BD Biosciences Supplier No. 341030
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CD55 antibody, clone IA10, is derived from the fusion of X63Ag8.653 myeloma cells with spleen cells from BALB/c mice immunized with purified DAF.

The CD55 antibody reacts with decay-accelerating factor (DAF), a GPI-anchored single chain glycoprotein of approximately 70 kilodaltons (kDa). It has been suggested that the role of DAF is to protect cells from damage by autologous complement preventing the amplification steps of the complement cascade by interfering with the assembly of the C3 and the C5 convertases. The complement components in the formation of C3 convertases include: C4b2a (classical pathway) and C3bBb (alternative pathway). In the formation of C5 convertases, the complement components include: C4b2a3b (classical pathway) and C3bBb3b (alternative pathway).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD55
Applications Flow Cytometry
Classification Monoclonal
Clone IA10
Concentration 25μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.