Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD55 Recombinant Rabbit Monoclonal Antibody (076), FITC
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA540932
This item is not returnable.
View return policy
Description
May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.
CD55 (decay-accelerating factor, DAF) is a GPI-anchored membrane glycoprotein that protects autologous cells from classical and alternative pathway of complement cascade. Bidirectional interactions between CD55 and CD97 are involved in T cell regulation and CD55 can regulate complement when bound to CD97. In tumors, CD55 promotes neoangiogenesis, tumorigenesis, invasiveness and evasion of apoptosis. CD55 is a 70 kDa glycoprotein (in erythrocytes) anchored in the membrane by glycosylphosphatidylinositol tail. In other cells, the apparent molecular weight is somewhat larger for CD55 and has a substantial content of O-glycans. CD55 binds to activated C4b or C3b complement fragments on the cell surface, preventing the assembly and accelerating the decay of both classical and alternative pathways. CD55 carries the Cromer related blood group antigens. Two alternatively spliced transcripts encoding CD55 have been identified, and the predominant transcript encodes a membrane-bound protein expressed on cells exposed to plasma component proteins but an alternatively spliced transcript produces a soluble protein present at much lower levels. CD55 has been studied to have a role in prostate cancer growth and survival.
Specifications
CD55 | |
Recombinant Monoclonal | |
0.1 mg/mL | |
PBS with 0.5% BSA and 0.09% sodium azide | |
Q61475 | |
CD55 | |
Recombinant Mouse CD55/DAF protein. | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry | |
076 | |
FITC | |
CD55 | |
CD55; CD55 antigen; CD55 molecule (Cromer blood group); CD55 molecule, decay accelerating factor for complement; CD55 molecule, decay accelerating factor for complement (Cromer blood group); Cd55a; Complement decay-accelerating factor; complement decay-accelerating factor, GPI-anchored; complement-glycosylphosphatidylinositol; CR; CROM; Cromer blood group; Daf; Daf1; DAF-GPI; decay accelarating factor 1; decay accelerating factor 1; decay accelerating factor GPI-form; decay-accelarating factor; glycosylphophatidylinositol-anchored form; GPI anchor addition signal; GPI-DAF; RP11-357P18.1; TC | |
Rabbit | |
Protein A | |
RUO | |
13136 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction