Learn More
Invitrogen™ CD56 Monoclonal Antibody (123C3)

Description
The antibody recognizes a transmembrane glycoprotein of 140 and 180 kD which has been identified as NCAM (Neural Cell Adhesion Module). At the international Workshop on SCLC antibodies 123C3 has been categorized as cluster 1 antibody. All cells in small cell carcinomas and carcinoids of the lung are strongly positive for 123C3. A minority of cases of other major types of lung carcinoma are sometimes positive as well: however this positivity is generally weak and focal. Adenoid cystic carcinomas of bronchial glands are strongly positive. Neuroblastoma's and Wilms tumors are usually also staining strongly positive. In non-small lung cell carcinomas, 123C3 staining has been associated with more advanced stage and a decreased survival after surgery. Furthermore, this antibody can be used to support diagnosis of lymphoma or to detect residual disease for cases of CD56 positive T/NK -cell lymphoma in which the neoplastic lymphoid cells are small and show minimal atypia, especially in small biopsies.
Specifications
Specifications
| Antigen | CD56 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 123C3 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide |
| Gene | Ncam1 |
| Gene Accession No. | P13591 |
| Gene Alias | adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.