Learn More
CD56 Monoclonal Antibody (C5.9), PerCP, Exalpha

Mouse Monoclonal Antibody
Supplier: Vector Laboratories 0565100TESTS


Description
Flow Cytometry PBMC: Add 10 μL of MAB/10^6 PBMC in 100 μL PBS. Mix gently and incubate for 15 minutes at 2 to 8C. Wash twice with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. Flow Cytometry Whole Blood: Add10 μL of MAB/100 μL of whole blood. Mix gently and incubate for 15 minutes at room temperature 20C. Lyse the whole blood. Wash once with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. See instrument manufacturer instructions for Lysed Whole Blood and Immunofluorescence analysis with a flow cytometer or microscope.
CD56 Monoclonal antibody specifically detects CD56 in Human samples. It is validated for Flow CytometrySpecifications
CD56 | |
Monoclonal | |
PerCP | |
Ncam1 | |
adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
Extract from KG1a cell line. | |
100Tests | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry | |
C5.9 | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P13591 | |
Mouse | |
Protein A/G | |
RUO | |
4684 | |
4° C, do not freeze | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.