Learn More
CD56 Mouse anti-Human, Alexa Fluor 488, Clone: B159, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 557699
Description
The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications
CD56 | |
Monoclonal | |
Alexa Fluor 488 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human NK Cells | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
B159 | |
NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.