Learn More
CD56 Mouse anti-Human, Alexa Fluor 700, Clone: B159, BD
Mouse Monoclonal Antibody
$138.60 - $373.50
Specifications
Antigen | CD56 |
---|---|
Clone | B159 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.
Specifications
CD56 | |
0.2mg/mL | |
Monoclonal | |
Mouse | |
Human | |
Human NK Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
B159 | |
Flow Cytometry | |
Alexa Fluor 700 | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.