Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD56 Mouse anti-Human, BB700, Clone: B159, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566401
Description
CD56 Monoclonal antibody specifically detects CD56 in Human samples. It is validated for Flow Cytometry.
Specifications
CD56 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
25 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
B159 | |
Mouse | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction